Thermo Scientific Chemicals

Cyclohexane, ACS, 99+%, Thermo Scientific Chemicals

Catalog number: 022864.AP
500 mL, Each
Thermo Scientific Chemicals

Cyclohexane, ACS, 99+%, Thermo Scientific Chemicals

Catalog number: 022864.AP
500 mL, Each
Quantity
Catalog number: 022864.AP
also known as 022864-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-82-7
IUPAC Name
cyclohexane
Molecular Formula
C6H12
InChI Key
XDTMQSROBMDMFD-UHFFFAOYSA-N
SMILES
C1CCCCC1
Appearance (Color)
Clear colorless
Form
Liquid
Color (APHA)
≤10
Assay (GC)
≥99.0% (corrected for water)
Residue on Evaporation
≤0.002%

Description

Cyclohexane is used in environmental and GC analysis; for industrial production of adipic acid and caprolactam, which is a precursor of nylon and as a solvent, and to make nylon and paint remover.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text