Thermo Scientific Chemicals

Crystal Violet, ACS, 90+%, Thermo Scientific Chemicals

Catalog number: 022866.14
25 g, Each
Thermo Scientific Chemicals

Crystal Violet, ACS, 90+%, Thermo Scientific Chemicals

Catalog number: 022866.14
25 g, Each
Quantity
Catalog number: 022866.14
also known as 022866-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
548-62-9
IUPAC Name
tris[4-(dimethylamino)phenyl]methylium chloride
Molecular Formula
C25H30ClN3
InChI Key
ZXJXZNDDNMQXFV-UHFFFAOYSA-M
SMILES
[Cl-].CN(C)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C
Appearance (Color)
Dark green to metallic green
Assay from Supplier's CofA
≥90.0%
Form
Powder or crystals or crystalline powder
Comment
Sensitivity as indicator P.T.
Comment
Absorbance characteristics P.T.

Description

Crystal Violet is used as an acid-base indicator, an alcohol denaturant, a biological stain, a textile dye and an indicator for copper salts. It is a component of gram staining in microorganisms and cell lines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text