Thermo Scientific Chemicals

Mullite, Thermo Scientific Chemicals

Catalog number: 022881.36
500 g, Each
Thermo Scientific Chemicals

Mullite, Thermo Scientific Chemicals

Catalog number: 022881.36
500 g, Each
Quantity
Catalog number: 022881.36
also known as 022881-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1302-93-8
IUPAC Name
tris(oxo[(oxoalumanyl)oxy]alumane); bis(silanedione)
Molecular Formula
Al6O13Si2
InChI Key
KZHJGOXRZJKJNY-UHFFFAOYSA-N
SMILES
O=[Si]=O.O=[Si]=O.O=[Al]O[Al]=O.O=[Al]O[Al]=O.O=[Al]O[Al]=O
Appearance (Color)
White to light grey
Particle Size
-325 mesh
Form
powder

Description

Mullite is present in the form of needles in porcelain, it is used in high-mullite based products is in hot blast stove checker bricks, melting and firing processes, and is used as a refractory material, manufacture of a type of crucible, steel industry, electronic substrates and protective coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text