Thermo Scientific Chemicals

Nickel aluminum, 99.0% min (metals basis), Thermo Scientific Chemicals

Catalog number: 022890.30
250 g, Each
Thermo Scientific Chemicals

Nickel aluminum, 99.0% min (metals basis), Thermo Scientific Chemicals

Catalog number: 022890.30
250 g, Each
Quantity
Catalog number: 022890.30
also known as 022890-30
Price (USD)
Have Questions?

Chemical Identifiers

CAS
12003-78-0
IUPAC Name
aluminium nickel
Molecular Formula
AlNi
InChI Key
NPXOKRUENSOPAO-UHFFFAOYSA-N
SMILES
[Al].[Ni]
Total Metal Impurities
1% max.

Description

Nickel aluminum is used in strengthening constituent in high-temperature nickel-base superalloys, coating blades in gas turbines and jet engines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text