Thermo Scientific Chemicals

Titanium aluminide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 022895.22
100 g, Each
Thermo Scientific Chemicals

Titanium aluminide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 022895.22
100 g, Each
Quantity
Catalog number: 022895.22
also known as 022895-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12004-78-3
IUPAC Name
trialuminium titanium
Molecular Formula
Al3Ti
InChI Key
OQPDWFJSZHWILH-UHFFFAOYSA-N
SMILES
[Al].[Al].[Al].[Ti]
Total Metal Impurities
0.5% max.

Description

Titanium aluminide is used in aircraft turbine engines, exhaust valves and turbochargers for passenger vehicle engines, intermediate process for some mill products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text