Thermo Scientific Chemicals

Potassium periodate, ACS, 99.8%-100.3%, Thermo Scientific Chemicals

Catalog number: 022896.22
100 g, Each
Thermo Scientific Chemicals

Potassium periodate, ACS, 99.8%-100.3%, Thermo Scientific Chemicals

Catalog number: 022896.22
100 g, Each
Quantity
Catalog number: 022896.22
also known as 022896-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7790-21-8
IUPAC Name
potassium periodate
Molecular Formula
IKO4
InChI Key
FJVZDOGVDJCCCR-UHFFFAOYSA-M
SMILES
[K+].[O-][I](=O)(=O)=O
Assay (unspecified)
99.8-100.3% KIO₄
Impurity content
Other halogens (as Cl) 0.01%
Impurity content
Mn 1ppm

Description

For the colorimetric determination of Mn (oxidizes manganese compounds to permanganates), as oxidizing agentPotassium periodate is used as an oxidizing agent in organic synthesis and an analytical reagent for the determination of potassium and cerium. It is also used in colorimetric estimation of manganese. It reacts with ammonium cerium(IV) nitrate to prepare tris[trinitratocerium(IV)] paraperiodate which finds application as an oxidant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text