Thermo Scientific Chemicals

2,2,4-Trimethylpentane, HPLC Grade, 99.7+%, Thermo Scientific Chemicals

Catalog number: 022901.K2
1 L, Each
Thermo Scientific Chemicals

2,2,4-Trimethylpentane, HPLC Grade, 99.7+%, Thermo Scientific Chemicals

Catalog number: 022901.K2
1 L, Each
Quantity
Catalog number: 022901.K2
also known as 022901-K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
540-84-1
IUPAC Name
2,2,4-trimethylpentane
Molecular Formula
C8H18
InChI Key
NHTMVDHEPJAVLT-UHFFFAOYSA-N
SMILES
CC(C)CC(C)(C)C
Optical Absorbance at
205nm: ≤1.00 AU
Optical Absorbance at
230nm: ≤0.10 au
Form
Liquid
Comment
Sub-micron filtered
Water Content (Karl Fischer Titration)
≤0.02%

Description

2,2,4-Trimethylpentane is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text