Thermo Scientific Chemicals

Toluene, HPLC Grade, 99.7% min, Thermo Scientific Chemicals

Catalog number: 022903.M1
4 x 1 L, Each
Thermo Scientific Chemicals

Toluene, HPLC Grade, 99.7% min, Thermo Scientific Chemicals

Catalog number: 022903.M1
4 x 1 L, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 022903.M1
also known as 022903-M1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-88-3
IUPAC Name
toluene
Molecular Formula
C7H8
InChI Key
YXFVVABEGXRONW-UHFFFAOYSA-N
SMILES
CC1=CC=CC=C1
Water Content (Karl Fischer Titration)
≤0.03% [US] / ≤0.01% [UK]
Optical Absorbance at
288nm: ≤0.40 AU [US]
Comment
Pesticide residue analysis: Passes test [UK]
Appearance (Color)
Colorless
Form
Liquid

Description

Toluene is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text