Pyridine, HPLC Grade, 99.5+%, Thermo Scientific Chemicals
Pyridine, HPLC Grade, 99.5+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pyridine, HPLC Grade, 99.5+%, Thermo Scientific Chemicals

 
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
4 x 1 L
Catalog number 022905.K7
also known as 022905-K7
Price (USD)/ Each
499.00
-
Add to cart
Quantity:
4 L
Request bulk or custom format
Price (USD)/ Each
499.00
Add to cart
Pyridine, HPLC Grade, 99.5+%, Thermo Scientific Chemicals
Catalog number022905.K7
Price (USD)/ Each
499.00
-
Add to cart
Chemical Identifiers
CAS110-86-1
IUPAC Namepyridine
Molecular FormulaC5H5N
InChI KeyJUJWROOIHBZHMG-UHFFFAOYSA-N
SMILESC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Absorbance at400nm: ≤0.01 AU`
FormClear liquid
Color (APHA)≤10
Assay (GC)≥ 99.5%
Water Content (Karl Fischer Titration)≤0.05%
View more
Pyridine is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only