n-Pentane, HPLC Grade, 99% min, Thermo Scientific Chemicals
n-Pentane, HPLC Grade, 99% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Pentane, HPLC Grade, 99% min, Thermo Scientific Chemicals

Maximum level of impurities: Evaporation residue 3ppm, H2O 0.01%
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 x 1 L
4 L
4 x 4 L
Catalog number 022907.K2
also known as 022907-K2
Price (USD)/ Each
61.65
Online exclusive
68.10 
Save 6.45 (9%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
61.65
Online exclusive
68.10 
Save 6.45 (9%)
Add to cart
n-Pentane, HPLC Grade, 99% min, Thermo Scientific Chemicals
Catalog number022907.K2
Price (USD)/ Each
61.65
Online exclusive
68.10 
Save 6.45 (9%)
-
Add to cart
Chemical Identifiers
CAS109-66-0
IUPAC Namepentane
Molecular FormulaC5H12
InChI KeyOFBQJSOFQDEBGM-UHFFFAOYSA-N
SMILESCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
CommentSub micron filtered
Optical Absorbance at210nm: ≤0.5 AU
Assay (GC)≥99.9% ( As C5 Hydrocarbons)
Optical Absorbance at200nm: ≤1.0 AU
Optical Absorbance at245nm: ≤0.01 AU
View more
n-Pentane is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only