Thermo Scientific Chemicals

n-Pentane, HPLC Grade, 99% min, Thermo Scientific Chemicals

Catalog number: 022907.M1
4 x 1 L, Each
Thermo Scientific Chemicals

n-Pentane, HPLC Grade, 99% min, Thermo Scientific Chemicals

Catalog number: 022907.M1
4 x 1 L, Each
Quantity
Catalog number: 022907.M1
also known as 022907-M1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-66-0
IUPAC Name
pentane
Molecular Formula
C5H12
InChI Key
OFBQJSOFQDEBGM-UHFFFAOYSA-N
SMILES
CCCCC
Comment
Sub micron filtered
Optical Absorbance at
210nm: ≤0.5 AU
Assay (GC)
≥99.9% ( As C5 Hydrocarbons)
Optical Absorbance at
200nm: ≤1.0 AU
Optical Absorbance at
245nm: ≤0.01 AU

Description

n-Pentane is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text