Thermo Scientific Chemicals

n-Heptane, HPLC grade, 99+%, Thermo Scientific Chemicals

Catalog number: 022911.K7
4 L, Each
Thermo Scientific Chemicals

n-Heptane, HPLC grade, 99+%, Thermo Scientific Chemicals

Catalog number: 022911.K7
4 L, Each
Quantity
Catalog number: 022911.K7
also known as 022911-K7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
142-82-5
IUPAC Name
heptane
Molecular Formula
C7H16
InChI Key
IMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILES
CCCCCCC
Appearance (Color)
Clear colorless
Water Content (Karl Fischer Titration)
≤0.02% [US & UK]
Optical Absorbance at
220nm: ≤0.05AU [US] / ≥80% transmittance [UK]
Assay (GC)
≥99.0%
Residue on Evaporation
≤0.00030% (3.0ppm) [US & UK]

Description

n-Heptane is used in paints and coatings, as the rubber cement solvent and GC analysis, HPLC, spectrophotometry and environmental testing. Used as a primary reference standard in determining the knock rating of carburetor fuels and solvent especially for vegetable oils, low temperature thermo- meters, calibrations, and polymerization reaction medium. The hydroisomerization of n-heptane to form dibranched and tribranched products useful for producing high octane gasoline. water/AOT/n-heptane microemulsions plays an essential role for the synthesis, size control, and passivation of CdS nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text