Thermo Scientific Chemicals

1,4-Dioxane, HPLC Grade, 99% min, Thermo Scientific Chemicals

Catalog number: 022913.K7
4 L, Each
Thermo Scientific Chemicals

1,4-Dioxane, HPLC Grade, 99% min, Thermo Scientific Chemicals

Catalog number: 022913.K7
4 L, Each
Quantity
Catalog number: 022913.K7
also known as 022913-K7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
123-91-1
IUPAC Name
1,4-dioxane
Molecular Formula
C4H8O2
InChI Key
RYHBNJHYFVUHQT-UHFFFAOYSA-N
SMILES
C1COCCO1
Residual water
0.03%
Residue on Evaporation
5ppm

Description

1,4-Dioxane HPLC Grade is used as a mobile phase in high performance liquid chromatography and liquid chromatography coupled with mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text