Thermo Scientific™

Dimethyl sulfoxide, HPLC Grade, 99.9+%, Thermo Scientific Chemicals

Catalog number: 022914.M1
4 x 1 L, Each
Thermo Scientific™

Dimethyl sulfoxide, HPLC Grade, 99.9+%, Thermo Scientific Chemicals

Catalog number: 022914.M1
4 x 1 L, Each
Quantity
Catalog number: 022914.M1
also known as 022914-M1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
67-68-5
IUPAC Name
methanesulfinylmethane
Molecular Formula
C2H6OS
InChI Key
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
SMILES
CS(C)=O
Appearance (Color)
Clear colorless
Form
Liquid
Assay from Suppliers CofA
≥99.9%
Optical Absorbance at
268nm: ≤1.00 AU
Optical Absorbance at
275nm: ≤0.50 AU

Description

Dimethyl sulfoxide is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text