Thermo Scientific Chemicals

N,N-Dimethylacetamide, HPLC Grade, 99.5+%, Thermo Scientific Chemicals

Catalog number: 022916.M1
4 x 1 L, Each
Thermo Scientific Chemicals

N,N-Dimethylacetamide, HPLC Grade, 99.5+%, Thermo Scientific Chemicals

Catalog number: 022916.M1
4 x 1 L, Each
Quantity
Catalog number: 022916.M1
also known as 022916-M1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
127-19-5
IUPAC Name
N,N-dimethylacetamide
Molecular Formula
C4H9NO
InChI Key
FXHOOIRPVKKKFG-UHFFFAOYSA-N
SMILES
CN(C)C(C)=O
Optical Absorbance at
270nm: ≤1.00AU [US sourced material]
Optical Absorbance at
275nm: ≤0.700AU [UK sourced material]
Optical Absorbance at
285nm: ≤0.301 [UK sourced material]
Optical Absorbance at
290nm: ≤0.15AU [US sourced material]
Optical Absorbance at
310nm: ≤0.046 [UK sourced material]

Description

N,N-Dimethylacetamide is used in all analytical laboratories as a major component of mobile phase in High Performance Liquid Chromatography (HPLC) and in Liquid Chromatography - Mass Spectrometry (LC-MS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text