Thermo Scientific Chemicals

Dichloromethane, HPLC Grade, 99.7+%, stab. with amylene, Thermo Scientific Chemicals

Catalog number: 022917.AK
250 mL, Each
Thermo Scientific Chemicals

Dichloromethane, HPLC Grade, 99.7+%, stab. with amylene, Thermo Scientific Chemicals

Catalog number: 022917.AK
250 mL, Each
Quantity
Catalog number: 022917.AK
also known as 022917-AK
Price (USD)
Have Questions?

Chemical Identifiers

CAS
75-09-2
IUPAC Name
dichloromethane
Molecular Formula
CH2Cl2
InChI Key
YMWUJEATGCHHMB-UHFFFAOYSA-N
SMILES
ClCCl
Optical Absorbance at
240nm: 0.20AU, max
Optical Absorbance at
250nm: 0.05AU, max
Optical Absorbance at
340nm: 0.01AU, max
Residue on Evaporation
3ppm, max
Stabilizer
40 to 100 ppm amylenes

Description

Dichloromethane is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text