Thermo Scientific Chemicals

1,2-Dichloroethane, HPLC Grade, 99% min, Thermo Scientific Chemicals

Catalog number: 022918.K7
4 L, Each
Thermo Scientific Chemicals

1,2-Dichloroethane, HPLC Grade, 99% min, Thermo Scientific Chemicals

Catalog number: 022918.K7
4 L, Each
Quantity
Catalog number: 022918.K7
also known as 022918-K7
Price (USD)
Have Questions?

Chemical Identifiers

CAS
107-06-2
IUPAC Name
1,2-dichloroethane
Molecular Formula
C2H4Cl2
InChI Key
WSLDOOZREJYCGB-UHFFFAOYSA-N
SMILES
ClCCCl
Assay from Supplier's CofA
≥99.0%
Residual water
≤0.02%
Residue on Evaporation
≤10 ppm
Appearance (Color)
Clear colorless
Form
Liquid

Description

1,2-Dichloroethane is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text