Thermo Scientific Chemicals

Cyclohexane, HPLC Grade, 99% min, Thermo Scientific Chemicals

Catalog number: 022919.K7
4 L, Each
Thermo Scientific Chemicals

Cyclohexane, HPLC Grade, 99% min, Thermo Scientific Chemicals

Catalog number: 022919.K7
4 L, Each
Quantity
Catalog number: 022919.K7
also known as 022919-K7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-82-7
IUPAC Name
cyclohexane
Molecular Formula
C6H12
InChI Key
XDTMQSROBMDMFD-UHFFFAOYSA-N
SMILES
C1CCCCC1
Optical Absorbance at
254 nm: ≤0.01 AU
Residue on Evaporation
≤1ppm
Form
Liquid
Color (APHA)
≤10
Assay (GC)
≥99.0%

Description

Cyclohexane is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text