Chloroform, HPLC Grade, 99.5+% min, Thermo Scientific Chemicals
Chloroform, HPLC Grade, 99.5+% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chloroform, HPLC Grade, 99.5+% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
4 x 1 L
1 L
4 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 022920.M1
also known as 022920-M1
Price (USD)/ Each
219.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
258.00 
Save 38.35 (15%)
-
Add to cart
Quantity:
4 x 1 L
Request bulk or custom format
Price (USD)/ Each
219.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
258.00 
Save 38.35 (15%)
Add to cart
Chloroform, HPLC Grade, 99.5+% min, Thermo Scientific Chemicals
Catalog number022920.M1
Price (USD)/ Each
219.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
258.00 
Save 38.35 (15%)
-
Add to cart
Chemical Identifiers
CAS67-66-3
IUPAC Nametrichloromethane
Molecular FormulaCHCl3
InChI KeyHEDRZPFGACZZDS-UHFFFAOYSA-N
SMILESClC(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤ 0.03%
Optical Absorbance at245 nm: ≤ 1.00 AU
Optical Absorbance at255 nm: ≤ 0.15 AU
Optical Absorbance at260 nm: ≤ 0.05 AU
Optical Absorbance at270 nm: ≤ 0.02 AU
View more
Chloroform is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only