Chlorobenzene, HPLC Grade, 99.5%, Thermo Scientific Chemicals
Chlorobenzene, HPLC Grade, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chlorobenzene, HPLC Grade, 99.5%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 x 1 L
4 L
Catalog number 022922.K2
also known as 022922-K2
Price (USD)/ Each
85.65
Online exclusive
95.50 
Save 9.85 (10%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
85.65
Online exclusive
95.50 
Save 9.85 (10%)
Add to cart
Chlorobenzene, HPLC Grade, 99.5%, Thermo Scientific Chemicals
Catalog number022922.K2
Price (USD)/ Each
85.65
Online exclusive
95.50 
Save 9.85 (10%)
-
Add to cart
Chemical Identifiers
CAS108-90-7
IUPAC Namechlorobenzene
Molecular FormulaC6H5Cl
InChI KeyMVPPADPHJFYWMZ-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Residual water0.03%
Residue on Evaporation2ppm
Chlorobenzene is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only