Thermo Scientific Chemicals

1-Butanol, HPLC Grade, 99%, Thermo Scientific Chemicals

Catalog number: 022925.K7
4 L, Each
Thermo Scientific Chemicals

1-Butanol, HPLC Grade, 99%, Thermo Scientific Chemicals

Catalog number: 022925.K7
4 L, Each
Quantity
Catalog number: 022925.K7
also known as 022925-K7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
71-36-3
IUPAC Name
butan-1-ol
Molecular Formula
C4H10O
InChI Key
LRHPLDYGYMQRHN-UHFFFAOYSA-N
SMILES
CCCCO
Assay (unspecified)
≥99%
Comment
Specification differs for U.S. and non-U.S. material where indicated
Comment
Submicron filtered
Optical Absorbance at
215 nm: ≤1.00 AU [U.S. specification]
Optical Absorbance at
220 nm: ≤0.50 AU [U.S. specification]

Description

1-Butanol is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text