Thermo Scientific™

Acetone, HPLC Grade, 99.5+%, Thermo Scientific Chemicals

Catalog number: 022928.M1
4 x 1 L, Each
Thermo Scientific™

Acetone, HPLC Grade, 99.5+%, Thermo Scientific Chemicals

Catalog number: 022928.M1
4 x 1 L, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 022928.M1
also known as 022928-M1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
67-64-1
IUPAC Name
propan-2-one
Molecular Formula
C3H6O
InChI Key
CSCPPACGZOOCGX-UHFFFAOYSA-N
SMILES
CC(C)=O
Comment
Sub micron filtered [US]
Impurity content
Fluorescence at Emission maximum for impurities ≤1ppb (as Quinine sulfate) [US]
Impurity content
Heptachlor epoxide: ≤10 ppt by GC-ECD [US]
Water Content (Karl Fischer Titration)
≤0.50% [US & UK]
Optical Absorbance at
329nm ≥10% transmittance [UK]

Description

Acetone, HPLC Grade is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry. It is useful as a laboratory reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text