Ethanol, anhydrous, denatured, HPLC Grade, 90%, 5% methanol, 5% isopropanol, Thermo Scientific Chemicals
Ethanol, anhydrous, denatured, HPLC Grade, 90%, 5% methanol, 5% isopropanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethanol, anhydrous, denatured, HPLC Grade, 90%, 5% methanol, 5% isopropanol, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
4 x 4 L
1 L
4 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 022930.M6
also known as 022930-M6
Price (USD)/ Each
353.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
416.00 
Save 62.35 (15%)
-
Add to cart
Quantity:
4 x 4 L
Request bulk or custom format
Price (USD)/ Each
353.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
416.00 
Save 62.35 (15%)
Add to cart
Ethanol, anhydrous, denatured, HPLC Grade, 90%, 5% methanol, 5% isopropanol, Thermo Scientific Chemicals
Catalog number022930.M6
Price (USD)/ Each
353.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
416.00 
Save 62.35 (15%)
-
Add to cart
Chemical Identifiers
CAS64-17-5
IUPAC Nameethanol
Molecular FormulaC2H6O
InChI KeyLFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILESCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Absorbance at230nm: 0.20AU, max;
Optical Absorbance at250nm: 0.05AU, max.
Optical Absorbance atat 270 nm 0.01AU, max. 210 nm 1.00 AU
Residue on Evaporation5ppm, max
Appearance (Color)Colorless
View more
Ethanol, anhydrous, denatured is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Manufacturing and Laboratory Use. Not for beverage use