Thermo Scientific Chemicals

Ethanol, anhydrous, denatured, Spectrophotometric Grade, 90%, 5% methanol, 5% isopropanol, Thermo Scientific Chemicals

Catalog number: 022931.K7
4 L, Each
Thermo Scientific Chemicals

Ethanol, anhydrous, denatured, Spectrophotometric Grade, 90%, 5% methanol, 5% isopropanol, Thermo Scientific Chemicals

Catalog number: 022931.K7
4 L, Each
Quantity
Catalog number: 022931.K7
also known as 022931-K7
Price (USD)
Have Questions?

Chemical Identifiers

CAS
64-17-5
IUPAC Name
ethanol
Molecular Formula
C2H6O
InChI Key
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES
CCO
Appearance (Color)
Colorless
Color (APHA)
10, max
Assay (GC)
4 to 6% 2-Propanol
Assay (GC)
4 to 6% Methanol
Assay (GC)
88 to 92% Ethanol

Description

Ethanol, anhydrous, denatured is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Manufacturing and Laboratory Use. Not for beverage use

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text