Thermo Scientific Chemicals

Water, ultrapure, HPLC Grade, Thermo Scientific Chemicals

Catalog number: 022934.M6
4 x 4 L, Each
Thermo Scientific Chemicals

Water, ultrapure, HPLC Grade, Thermo Scientific Chemicals

Catalog number: 022934.M6
4 x 4 L, Each
Quantity
Catalog number: 022934.M6
also known as 022934-M6
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7732-18-5
IUPAC Name
water
Molecular Formula
H2O
InChI Key
XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES
O
Appearance (Color)
Clear colorless
Assay (unspecified)
Specific Conductance at 25 deg C and time packaging: 1.5 microS/cm, max
Optical Absorbance at
400 nm 0.005 AU, max- at 300 nm 0.005 AU, max- at 250 nm 0.005 AU, max- at 200 nm 0.010 AU, max- at 190 nm 0.010 AU, max
Residue on Evaporation
2 ppm, Max
Form
Liquid

Description

Water ultrapure is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry. It is also used in conductometric titration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text