Thermo Scientific Chemicals

Vanadium(III) 2,4-pentanedionate, typically 97%, Thermo Scientific Chemicals

Catalog number: 022940.06
5 g, Each
Thermo Scientific Chemicals

Vanadium(III) 2,4-pentanedionate, typically 97%, Thermo Scientific Chemicals

Catalog number: 022940.06
5 g, Each
Quantity
Catalog number: 022940.06
also known as 022940-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13476-99-8
IUPAC Name
vanadium(3+) tris(2,4-dioxopentan-3-ide)
Molecular Formula
C15H21O6V
InChI Key
ZVFSRLXFKIWGRT-UHFFFAOYSA-N
SMILES
[V+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Appearance (Color)
Green to brown
Assay (unspecified)
Vanadium (V): ≥14.0% (≥ 96.0% purity calc.)
Form
Powder

Description

Vanadium(III) 2,4-pentanedionate is used as homogeneous catalyst, primary and secondary intermediates, Laboratory chemicals, and manufacture of substances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text