Thermo Scientific Chemicals

Dimethyl sulfide, 99+%, Thermo Scientific Chemicals

Catalog number: 022949.AK
250 mL, Each
Thermo Scientific Chemicals

Dimethyl sulfide, 99+%, Thermo Scientific Chemicals

Catalog number: 022949.AK
250 mL, Each
Quantity
Catalog number: 022949.AK
also known as 022949-AK
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-18-3
IUPAC Name
(methylsulfanyl)methane
Molecular Formula
C2H6S
InChI Key
QMMFVYPAHWMCMS-UHFFFAOYSA-N
SMILES
CSC
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥99.0%
Refractive Index
1.4260-1.4460 @ 20?C

Description

Dimethyl sulfide is used as a displaceable ligand in chloro(dimethyl sulfide)gold(I) and other coordination compounds, as a reducing agent in ozonolysis reactions as well as a food flavoring component. Further, it is utilized for the preparation of dimethyl sulfoxide, which is a common and important industrial solvent. It finds application in the petroleum industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text