Dimethyl sulfide, 99+%, Thermo Scientific Chemicals
Dimethyl sulfide, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethyl sulfide, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Catalog number 022949.K2
also known as 022949-K2
Price (USD)/ Each
58.65
Online exclusive
65.10 
Save 6.45 (10%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
58.65
Online exclusive
65.10 
Save 6.45 (10%)
Add to cart
Dimethyl sulfide, 99+%, Thermo Scientific Chemicals
Catalog number022949.K2
Price (USD)/ Each
58.65
Online exclusive
65.10 
Save 6.45 (10%)
-
Add to cart
Chemical Identifiers
CAS75-18-3
IUPAC Name(methylsulfanyl)methane
Molecular FormulaC2H6S
InChI KeyQMMFVYPAHWMCMS-UHFFFAOYSA-N
SMILESCSC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥99.0%
Refractive Index1.4260-1.4460 @ 20?C
Dimethyl sulfide is used as a displaceable ligand in chloro(dimethyl sulfide)gold(I) and other coordination compounds, as a reducing agent in ozonolysis reactions as well as a food flavoring component. Further, it is utilized for the preparation of dimethyl sulfoxide, which is a common and important industrial solvent. It finds application in the petroleum industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only