Thermo Scientific Chemicals

Iron boride, 98% (metals basis), Thermo Scientific Chemicals

Catalog number: 022952.14
25 g, Each
Thermo Scientific Chemicals

Iron boride, 98% (metals basis), Thermo Scientific Chemicals

Catalog number: 022952.14
25 g, Each
Quantity
Catalog number: 022952.14
also known as 022952-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12006-85-8
IUPAC Name
boron diiron
Molecular Formula
BFe2
InChI Key
FSDZRQFSRALZQJ-UHFFFAOYSA-N
SMILES
[B].[Fe].[Fe]
Form
Powder
Total Metal Impurities
≤2.0%

Description

Iron boride is used in the production of borated steels and borated stainless steels, deoxidizing agents, increase the hardness of steel, and alloying components of coatings applied to weld-facing electrodes. It is also used to improve abrasion resistance, wear resistance, and oxidation resistance. It is used in oil and gas refinery, chemical extraction, automotive, agricultural, stamping, textile extrusion and injection molding industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text