Vanadium(IV) oxide, 99% (metals basis), Thermo Scientific Chemicals
Vanadium(IV) oxide, 99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Vanadium(IV) oxide, 99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 022957.09
also known as 022957-09
Price (USD)/ Each
80.65
Online exclusive
89.90 
Save 9.25 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
80.65
Online exclusive
89.90 
Save 9.25 (10%)
Add to cart
Vanadium(IV) oxide, 99% (metals basis), Thermo Scientific Chemicals
Catalog number022957.09
Price (USD)/ Each
80.65
Online exclusive
89.90 
Save 9.25 (10%)
-
Add to cart
Chemical Identifiers
CAS12036-21-4
IUPAC Namedioxovanadium
Molecular FormulaO2V
InChI KeyGRUMUEUJTSXQOI-UHFFFAOYSA-N
SMILESO=[V]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
Vanadium(IV) oxide is used in chemical sensors, energy-conserving coatings, transparent conductors and switching materials. It acts as a stationary optical shutter, optical modulators, cameras and data storage. As infrared modulators, it is used for the missile guidance systems. Thin films made up of vanadium dioxide find applications in electro-optical switches, micro-optical switch, passive smart radiators and sunshields for spacecraft.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only