Thermo Scientific Chemicals

Vanadium(IV) oxide, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 022957.18
50 g, Each
Thermo Scientific Chemicals

Vanadium(IV) oxide, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 022957.18
50 g, Each
Quantity
Catalog number: 022957.18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12036-21-4
IUPAC Name
dioxovanadium
Molecular Formula
O2V
InChI Key
GRUMUEUJTSXQOI-UHFFFAOYSA-N
SMILES
O=[V]=O
Total Metal Impurities
1% max.

Description

Vanadium(IV) oxide is used in chemical sensors, energy-conserving coatings, transparent conductors and switching materials. It acts as a stationary optical shutter, optical modulators, cameras and data storage. As infrared modulators, it is used for the missile guidance systems. Thin films made up of vanadium dioxide find applications in electro-optical switches, micro-optical switch, passive smart radiators and sunshields for spacecraft.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text