Molybdic acid, ACS, MoO{3} 85% min, Thermo Scientific Chemicals
Molybdic acid, ACS, MoO{3} 85% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Molybdic acid, ACS, MoO{3} 85% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
500 g
Catalog number 022983.A3
also known as 022983-A3
Price (USD)/ Each
540.65
Online exclusive
601.00 
Save 60.35 (10%)
-
Add to cart
Quantity:
2 kg
Request bulk or custom format
Price (USD)/ Each
540.65
Online exclusive
601.00 
Save 60.35 (10%)
Add to cart
Molybdic acid, ACS, MoO{3} 85% min, Thermo Scientific Chemicals
Catalog number022983.A3
Price (USD)/ Each
540.65
Online exclusive
601.00 
Save 60.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7782-91-4
IUPAC Namedioxomolybdenumdiol
Molecular FormulaH2MoO4
InChI KeyVLAPMBHFAWRUQP-UHFFFAOYSA-L
SMILESO[Mo](O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentPO₄ 5ppm
Assay (unspecified)85.0% MoO₃ min
Impurity contentArsenate, phosphate, and silicate (as SiO₂) 0.001%
Impurity contentCl 0.002%
Impurity contentInsoluble in dilute ammonium hydroxide 0.01%
View more
Molybdic acid is used as a precursor in the production of ammonium molybdate tetrahydrate and molybdenum(IV) sulfide. The mixture of molybdic acid in concentrated sulfuric acid is called as Froehde reagent, which finds application in analytical chemistry for the detection of alkaloids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only