Thermo Scientific Chemicals

Phosphorus(III) chloride, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog number: 022988.14
25 g, Each
Thermo Scientific Chemicals

Phosphorus(III) chloride, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog number: 022988.14
25 g, Each
Quantity
Catalog number: 022988.14
also known as 022988-14
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7719-12-2
IUPAC Name
trichlorophosphane
Molecular Formula
Cl3P
InChI Key
FAIAAWCVCHQXDN-UHFFFAOYSA-N
SMILES
ClP(Cl)Cl
Total Metal Impurities
0.003% max.

Description

Phosphorus(III) chloride is the precursor to organ phosphorus compounds, most notably phosphites and phosphonates. It is used during electrode position of metal on rubber and for making surfactants, gasoline additives, dyestuffs, textile finishing agents, medicinal products, and other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text