Thermo Scientific Chemicals

Zirconium(IV) n-propoxide, 70% w/w in n-propanol, Thermo Scientific Chemicals

Catalog number: 022989.18
50 g, Each
Thermo Scientific Chemicals

Zirconium(IV) n-propoxide, 70% w/w in n-propanol, Thermo Scientific Chemicals

Catalog number: 022989.18
50 g, Each
Quantity
Catalog number: 022989.18
Price (USD)
Have Questions?

Chemical Identifiers

CAS
23519-77-9
IUPAC Name
tetrapropoxyzirconium
Molecular Formula
C12H28O4Zr
InChI Key
XPGAWFIWCWKDDL-UHFFFAOYSA-N
SMILES
CCCO[Zr](OCCC)(OCCC)OCCC
Appearance (Color)
Clear yellow to brown
Form
Liquid
Assay (unspecified)
ZrO2: 27.0 to 29.0%
Chloride content
≤500 ppm
Specific Gravity
1.05-1.09

Description

Zirconium(IV) n-propoxide is used as a precursor in the production of zirconia. It is also used to study the thermophysical properties like thermal conductivity and thermal effusivity of hybrid films and thereby adjusting optic properties of the films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text