Cobalt(II) acetate, anhydrous, Co 32% min., Thermo Scientific Chemicals
Cobalt(II) acetate, anhydrous, Co 32% min., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cobalt(II) acetate, anhydrous, Co 32% min., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number 023138.22
also known as 023138-22
Price (USD)/ Each
722.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
722.00
Add to cart
Cobalt(II) acetate, anhydrous, Co 32% min., Thermo Scientific Chemicals
Catalog number023138.22
Price (USD)/ Each
722.00
-
Add to cart
Chemical Identifiers
CAS71-48-7
IUPAC Nameλ²-cobalt(2+) diacetate
Molecular FormulaC4H6CoO4
InChI KeyQAHREYKOYSIQPH-UHFFFAOYSA-L
SMILES[Co++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)96% min
Cobalt(II) acetate is used as an industrial catalyst. It is used as a precursor to various oil drying agents. It finds application in ion exchange agents, lubricants, plating agents and surface treating agents, greases, ink, toner, and colorant products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only