Cobalt(II) oxide, 95%, Thermo Scientific Chemicals
Cobalt(II) oxide, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cobalt(II) oxide, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number 023141.22
also known as 023141-22
Price (USD)/ Each
309.65
Online exclusive
344.00 
Save 34.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
309.65
Online exclusive
344.00 
Save 34.35 (10%)
Add to cart
Cobalt(II) oxide, 95%, Thermo Scientific Chemicals
Catalog number023141.22
Price (USD)/ Each
309.65
Online exclusive
344.00 
Save 34.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1307-96-6
IUPAC Nameoxocobalt
Molecular FormulaCoO
InChI KeyIVMYJDGYRUAWML-UHFFFAOYSA-N
SMILESO=[Co]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)94.1% min

Gallium(III) oxide is used in vacuum deposition. It is useful for making semiconductor devices, gallium-alumina catalyst, gas sensors, luminescent phosphors and dielectric coatings of solar cells. It shows potential for developing deep-ultraviolet TCOs (Transparent Conductive Oxides) and transparent electrodes for ultraviolet optoelectronic devices. Recent studies report that gallium oxide can be a strong contender for power electronic devices for example, in ultrahigh-voltage power switching applications. Films made of gallium oxide have gained commercial interest owing to their gas sensitive characteristics, and glasses made with gallium oxide are the preferred optical materials for use in advanced technologies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only