Thermo Scientific Chemicals

Cobalt(II) bromide, anhydrous, 97%, Thermo Scientific Chemicals

Catalog number: 023149.18
50 g, Each
Thermo Scientific Chemicals

Cobalt(II) bromide, anhydrous, 97%, Thermo Scientific Chemicals

Catalog number: 023149.18
50 g, Each
Quantity
Catalog number: 023149.18
also known as 023149-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7789-43-7
IUPAC Name
λ²-cobalt(2+) dibromide
Molecular Formula
Br2Co
InChI Key
BZRRQSJJPUGBAA-UHFFFAOYSA-L
SMILES
[Co++].[Br-].[Br-]
Assay (unspecified)
96.1% min

Description

Cobalt(II) bromide is used as a catalyst in organic synthesis. It is used as a precursor in the production of ethylsulfanyl)porphyrazinato)cobalt(II), which gives the possibility of intermolecular ferromagnetic interactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text