Tricarbonylnitrosylcobalt, Thermo Scientific Chemicals
Tricarbonylnitrosylcobalt, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tricarbonylnitrosylcobalt, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 023151.06
also known as 023151-06
Price (USD)/ Each
297.65
Online exclusive
330.00 
Save 32.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
297.65
Online exclusive
330.00 
Save 32.35 (10%)
Add to cart
Tricarbonylnitrosylcobalt, Thermo Scientific Chemicals
Catalog number023151.06
Price (USD)/ Each
297.65
Online exclusive
330.00 
Save 32.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialTricarbonylnitrosylcobalt
CAS14096-82-3
Health Hazard 1H225-H301-H330-H334-H350
Health Hazard 2GHS H Statement
H225-H301-H330-H351-H317
Highly flammable liquid and vapor.
Toxic if swallowed.
Fatal if inhaled.
Suspected of causing cancer.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P284-P285-P301+P310-P303+P361+P353-P304+P340-P308+P313-P310-P330-P370+P378q-P501c
View more
Tricarbonylnitrosylcobalt is used as the Co metalorganic precursor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only