Chromate, Quant™ Test Strips, Thermo Scientific Chemicals
Chromate, Quant™ Test Strips, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chromate, Quant™ Test Strips, Thermo Scientific Chemicals
Have Questions?
Quantity:
1 Kit
Catalog number 023274.QA
also known as 023274-QA
Price (USD)/ Each
254.00
-
Add to cart
Quantity:
1 Kit
Request bulk or custom format
Price (USD)/ Each
254.00
Add to cart
Chromate, Quant™ Test Strips, Thermo Scientific Chemicals
Catalog number023274.QA
Price (USD)/ Each
254.00
-
Add to cart
Specifications
Quantity1 Kit
RangeMeasures 3 to 1000ppm CrO4-2 in aqueous solution
TypeChromate test strip
Unit SizeEach
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only