Formaldehyde, Quant™ Test Strips, Thermo Scientific Chemicals
Formaldehyde, Quant™ Test Strips, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Formaldehyde, Quant™ Test Strips, Thermo Scientific Chemicals
Have Questions?
Quantity:
1 Kit
Catalog number 023277.QA
also known as 023277-QA
Price (USD)/ Each
238.00
-
Add to cart
Quantity:
1 Kit
Request bulk or custom format
Price (USD)/ Each
238.00
Add to cart
Formaldehyde, Quant™ Test Strips, Thermo Scientific Chemicals
Catalog number023277.QA
Price (USD)/ Each
238.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Quantity1 Kit
RangeMeasures 0 to 100ppm HCHO in aqueous solution
TypeFormaldehyde test strip