Sulfate, Quantofix™ Test Strips, Thermo Scientific Chemicals
Catalog number023287.QA
Price (USD)/ Each
98.20
-
Add to cart
Specifications
Chemical Name or MaterialSulfate, Test strips
Recommended StorageAmbient temperatures
Quantity1 Kit
TSCANo
MDL NumberMFCD00148535
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.