Chromium(III) phosphate tetrahydrate, Thermo Scientific Chemicals
Chromium(III) phosphate tetrahydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chromium(III) phosphate tetrahydrate, Thermo Scientific Chemicals

For Use As a Catalyst, Pigment
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 024110.09
also known as 024110-09
Price (USD)/ Each
43.50
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
43.50
Add to cart
Chromium(III) phosphate tetrahydrate, Thermo Scientific Chemicals
Catalog number024110.09
Price (USD)/ Each
43.50
-
Add to cart
Chemical Identifiers
CAS10101-59-4
IUPAC Namechromium(3+) tetrahydrate phosphate
Molecular FormulaCrH8O8P
InChI KeyBVZHAJJRIRDSTG-UHFFFAOYSA-K
SMILESO.O.O.O.[Cr+3].[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97% min
It is used as an additive in drugs, anti-corrosive pigments, catalysts and polymer manufacture, is also used to catalyze cation exchange in sorption reactions, as a laminated structure to dampen vibration and noise in a motor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only