Specifications
Chemical Name or MaterialTris(ethylenediamine)chromium(III) chloride hemiheptahydrate
CAS16165-32-5
Health Hazard 1H317
Health Hazard 2GHS H Statement
H317
May cause an allergic skin reaction.
Health Hazard 3P261-P272-P280g-P302+P352-P333+P313-P363-P501c
View more