Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Description | Style | Type |
---|---|---|---|
02PINK also known as 02 PINK | Pink | Vial | |
0385 | 90mL (3 oz.); With skirt top | Skirt top | Polypropylene Container |
03CLFREGORED | 3 oz. (90mL); Frego Red | Standard | Polypropylene Container |
03EDM33 | 90mL (3 oz.); With write-on space | Write-on space | Polypropylene Container |
04HP-SMOOTH | 4 oz. (118mL); High profile; With write-on space | High profile with write-on space | Polypropylene Container |
04LP | 4 oz. (118mL); Low profile; Natural | Low profile | Polypropylene Container |
05WIDE | 5 oz. (148mL); Wide mouth; Natural | Low profile, wide mouth | Polypropylene Container |
08CL | 8 oz. (236mL); Natural | Standard | Polypropylene Container |
2.5CL | 2.5 oz. (74mL); Natural | Standard | Polypropylene Container |
2.5EVERGREEN | 2.5 oz. (74mL); Evergreen | Standard | Polypropylene Container |
2.5RED | 2.5 oz. (74mL); Red | Standard | Polypropylene Container |
02CL | 1.5 oz. (45mL); Natural | Polypropylene Container | |
02DKORANGE also known as 02 DKORANGE | Vial | ||
02EVERGREEN also known as 02 EVERGREEN | Vial | ||
02FREGORED also known as 02 FREGORED | Vial | ||
02LIME also known as 02 LIME | Vial | ||
02ROYALBLUE also known as 02 ROYALBLUE | Royal Blue | Vial | |
02SKYBLUE also known as 02 SKYBLUE | Sky Blue | Vial | |
02YELLOW also known as 02 YELLOW | Vial | ||
02TURQUOISE also known as 02 TURQUOISE | Vial | ||
02PURPLE also known as 02 PURPLE | Vial |
Thermo Scientific™ Capitol Vial Flip-Top Polypropylene Containers are ideal for collecting liquids and solids for dairy testing, food and beverage sample collection, veterinary applications, environmental sampling, water analysis, general packaging and storag.
Recommended for:
Dairy testing, food and beverage sample collection, veterinary applications, environmental sampling, water testing, general packaging and storage