Al-23 Tube, One End Closed, O.D.: 4mm, I.D.: 2mm, Thermo Scientific Chemicals
Al-23 Tube, One End Closed, O.D.: 4mm, I.D.: 2mm, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Al-23 Tube, One End Closed, O.D.: 4mm, I.D.: 2mm, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
600 mm
200 mm
300 mm
800 mm
1000 mm
Catalog number 030062.EH
also known as 030062-EH
Price (USD)/ Each
82.50
-
Add to cart
Quantity:
600 mm
Request bulk or custom format
Price (USD)/ Each
82.50
Add to cart
Al-23 Tube, One End Closed, O.D.: 4mm, I.D.: 2mm, Thermo Scientific Chemicals
Catalog number030062.EH
Price (USD)/ Each
82.50
-
Add to cart
Specifications
Chemical Name or MaterialAl-23 Tube
Name NoteOne end closed
Recommended StorageAmbient temperatures
TSCAU
Quantity600 mm