Thermo Scientific Chemicals

Ammonium O,O'-diethyl dithiophosphate, typically 95%, Thermo Scientific Chemicals

Catalog number: 030139.14
25 g, Each
Thermo Scientific Chemicals

Ammonium O,O'-diethyl dithiophosphate, typically 95%, Thermo Scientific Chemicals

Catalog number: 030139.14
25 g, Each
Quantity
Catalog number: 030139.14
also known as 030139-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1068-22-0
IUPAC Name
ammonium O,O-diethyl sulfanidylphosphonothioate
Molecular Formula
C4H14NO2PS2
InChI Key
HFRHTRKMBOQLLL-UHFFFAOYSA-N
SMILES
[NH4+].CCOP([S-])(=S)OCC
Appearance (Color)
White
Form
Powder or crystals
Elemental Analysis
Carbon: 22.2 - 25.1 %
Elemental Analysis
Nitrogen: 6.4 - 7.3 %
Infrared spectrum
Conforms

Description

Diethyl dithiophosphate ammonium salt can be used in the extraction of arsenic. It is used as a source of the (C2H5O)2PS2- ligand in coordination chemistry and in analytical chemistry for determination of various ions. It can be obtained by the reaction of phosphorus pentasulfide with ethanol and ammonia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text