Thermo Scientific Chemicals
Trimethylphosphine, 98%, Thermo Scientific Chemicals
Catalog number: 030143.14
25 g, Each
Thermo Scientific Chemicals
Trimethylphosphine, 98%, Thermo Scientific Chemicals
Catalog number: 030143.14
25 g, Each
Quantity
Catalog number: 030143.14
also known as 030143-14
Price (USD)
Quantity
-
Have Questions?
Description
It is used in the preparation of organic phosphorus compounds and frequently-used phosphine ligand. It is also a highly basic ligand that forms complexes with most metals. In addition, it can react with bromomethyl-benzene to get benzyl-trimethyl-phosphonium; bromide. 