Thermo Scientific Chemicals

Phenylphosphine, Thermo Scientific Chemicals

Catalog number: 030155.06
Each
Thermo Scientific Chemicals

Phenylphosphine, Thermo Scientific Chemicals

Catalog number: 030155.06
Each
Catalog number: 030155.06
also known as 030155-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
638-21-1
IUPAC Name
phenylphosphane
Molecular Formula
C6H7P
InChI Key
RPGWZZNNEUHDAQ-UHFFFAOYSA-N
SMILES
PC1=CC=CC=C1
Assay (GC)
>98.5%
Proton NMR
Conforms to structure
Phosphorus-31 NMR
Conforms to structure
Appearance (Color)
Clear, colorless
Form
Liquid

Description

It is mainly used as a precursor to other organophosphorus compounds. It can function as a ligand in coordination chemistry. Phenylphosphine also have uses in polymer synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text