Thermo Scientific Chemicals

Tris(2-cyanoethyl)phosphine, 94%, Thermo Scientific Chemicals

Catalog number: 030165.09
10 g, Each
Thermo Scientific Chemicals

Tris(2-cyanoethyl)phosphine, 94%, Thermo Scientific Chemicals

Catalog number: 030165.09
10 g, Each
Quantity
Catalog number: 030165.09
also known as 030165-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4023-53-4
IUPAC Name
3-[bis(2-cyanoethyl)phosphanyl]propanenitrile
Molecular Formula
C9H12N3P
InChI Key
CHZAMJVESILJGH-UHFFFAOYSA-N
SMILES
N#CCCP(CCC#N)CCC#N
Form
Powder or crystalline powder
Appearance (Color)
White to yellow to pale brown
Assay by NMR
≥92.0% (Phosphorus NMR) (U.K. Specification)
Comment
Purchased in U.K. and U.S.
Phosphorus-31 NMR
Conforms to structure (U.K. Specification)

Description

Tris(2-cyanoethyl)phosphine is used to produce 3,3',3''-phosphanetriyl-tri-propionic acid and hydrochloride by heating. It will need reagent conc. aq. HCl with reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text