Thermo Scientific Chemicals

Dichloro[bis(1,3-diphenylphosphino)propane]nickel(II), Thermo Scientific Chemicals

Catalog number: 030167.09
10 g, Each
Thermo Scientific Chemicals

Dichloro[bis(1,3-diphenylphosphino)propane]nickel(II), Thermo Scientific Chemicals

Catalog number: 030167.09
10 g, Each
Quantity
Catalog number: 030167.09
also known as 030167-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15629-92-2
IUPAC Name
[3-(diphenylphosphanyl)propyl]diphenylphosphane; dichloronickelbis(ylium)
Molecular Formula
C27H26Cl2NiP2
InChI Key
REQVOKPBMSCTNI-UHFFFAOYSA-L
SMILES
Cl[Ni++]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
Orange to red
Elemental Analysis
% H: 4.7 - 4.9%
Elemental Analysis
%C: 59.2 - 60.5%
Form
powder

Description

Dichloro[bis(1,3-diphenylphosphino)propane]nickel(II) is used as an effective catalyst for Kumada coupling and Suzuki reactions. It also catalyzes other reactions to convert enol ethers, dithioacetals and vinyl sulfides to olefins. Further, it is used as a catalyst for preparation of block-copolythiophenes and solid state metathesis polycondensation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text